
2-Bromo-5-methoxyphenylboronic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:89694-44-0
:
Formula:C₇H₈BBrO₃
MDL Number:MFCD06659859
Melting Pt:214…216 °C
:230.85 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together