Order Entry
Portugal
ContactUsLinkComponent
Potassium bromide for IR spectrophotometry
Potassium bromide for IR spectrophotometry
Catalog # APOSIN2888-100G
Supplier:  Apollo Scientific
CAS Number:  
Potassium bromide for IR spectrophotometry
Catalog # APOSIN2888-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
Boiling Pt:1435 °C
Melting Pt:734 °C
Density:2.75 g/cm³ (25 °C)
 :119 g/mol
Storage Temperature:Ambient
EINECS:231-830-3
 :GHS07
CAS Number:7758-02-3
Formula:KBr
MDL Number:MFCD00011358