Order Entry
Portugal
ContactUsLinkComponent
Calcium carbonate 99.999%
Calcium carbonate 99.999%
Catalog # APOSIN1384-25G
Supplier:  Apollo Scientific
CAS Number:  
Calcium carbonate 99.999%
Catalog # APOSIN1384-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :100.09 g/mol
Melting Pt:800 °C (dec.)
Density:2.93 g/cm³ (25 °C)
EINECS:207-439-9
 :GHS07
Storage Temperature:Ambient
CAS Number:471-34-1
Formula:CaCO₃
MDL Number:MFCD00010906