Order Entry
Portugal
Orders LinkContactUsLinkComponent
Tween® 80 (Polysorbate)

Tween® 80 (Polysorbate)

Irritant
Supplier:   Apollo Scientific
Tween® 80 (Polysorbate)
Supplier:  Apollo Scientific
Certificates
Tween® 80 (Polysorbate)

3 Options Available Below

Product Details & Documents

Non ionic detergent used for cell lysis, nuclei isolationand cell fractionationBulk pricing available

  • CAS Number:9005-65-6
  • MDL Number:MFCD00082107
  • Boiling Pt:100 °C
  • Melting Pt:–25 °C
  • Density:1.064 g/cm³ (25 °C)
  • Flash Pt:>113 °C
  • Storage Temperature:Ambient
  • EINECS:500-019-9

Documents

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 55,00 €
    55,00 €
    1 * 1 L
       55,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 2 L
    1 * 2 L - 102,00 €
    102,00 €
    1 * 2 L
       102,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 232,00 €
    232,00 €
    1 * 5 L
       232,00 €

    Specifications:
    New Product
    New Product
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.