Order Entry
Portugal
ContactUsLinkComponent
APS-2-79 hydrochloride
APS-2-79 hydrochloride
Catalog # APOSBISN0199-5MG
Supplier:  Apollo Scientific
CAS Number:  
APS-2-79 hydrochloride
Catalog # APOSBISN0199-5MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :423.89 g/mol
Storage Temperature:Refrigerator
UN:3077
ADR:9+EHS,III
CAS Number:2002381-31-7
Formula:C₂₃H₂₂ClN₃O₃
MDL Number:MFCD30536203