Sorafenib (BAY 43-9006)
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:284461-73-0
:
Formula:C₂₁H₁₆ClF₃N₄O₃
MDL Number:MFCD06411450
:464.82 g/mol
Melting Pt:197 °C
Density:1.46 g/cm³ (25 °C)
Storage Temperature:Refrigerator
UN:3077
ADR:9+EHS,III
Frequently Bought Together