Hydroxy-PEG8-t-butyl ester
Catalog # APOSBIPG1454-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:498.60 g/mol
Storage Temperature:Ambient
CAS Number:1334177-84-2
Formula:C₂₃H₄₆O₁₁
MDL Number:MFCD11041105