Order Entry
Portugal
Orders LinkContactUsLinkComponent
 

Fluorescamine

Irritant
Supplier:   Apollo Scientific
Fluorescamine
Supplier:  Apollo Scientific
Certificates

4 Options Available Below

Product Details & Documents

Reacts with primary amines to form stable, highlyfluorescent products.

Synonyms:  Fluram, 4-Phenyl-3H,3'H-spiro[furan-2,1'-isobenzofuran]-3,3'-dione, 4-Phenylspiro[furan-2(3H),1'-phthalan]-3,3'-dione
  • CAS Number:38183-12-9
  • Formula:C₁₇H₁₀O₄
  • MDL Number:MFCD00005928
  • Molecular Weight:278.26 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 88,30 €
    88,30 €
    1 * 100 mg
       88,30 €

    Specifications:
    New Product
    New Product
  • Bottle
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 154,00 €
    154,00 €
    1 * 250 mg
       154,00 €

  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 313,00 €
    313,00 €
    1 * 1 g
       313,00 €

  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 1 100,00 €
    1 100,00 €
    1 * 5 g
       1 100,00 €