Chloramphenicol ≥99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:56-75-7
:
Formula:Cl₂CHCONHCH(CH₂OH)CH(OH)C₆H₄NO₂
MDL Number:MFCD00078159
:323.13 g/mol
Melting Pt:148…152 °C
EINECS:200-287-4
:GHS07
:GHS08
Storage Temperature:Refrigerator
Frequently Bought Together