
Phenyl-β-D-thioglucopyranoside
Catalog # APOSBIB1429-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:272.317 g/mol
:GHS07
Storage Temperature:Freezer
CAS Number:2936-70-1
Formula:C₁₂H₁₆O₅S
MDL Number:MFCD11045365