Methyl 3-nitro-4-[3-(trifluoromethyl)phenoxy]benzoate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:320417-31-0
:
Formula:C₁₅H₁₀F₃NO₅
MDL Number:MFCD00138767
:341.239 g/mol
Melting Pt:74…76 °C
Storage Temperature:Ambient
Frequently Bought Together