(L)-4,4,4,-Trifluoro-α-homoalanine, N-FMOC protected
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4,4,4-trifluorobutanoic acid, (L)-2-Amino-4,4,4-trifluorobutanoic acid, N-FMOC protected
CAS Number:181128-48-3
:
Formula:C₁₉H₁₆F₃NO₄
MDL Number:MFCD08532485
:379.33 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together