
2,2,2-Trifluoro-1-(4-fluorophenyl)ethylamine hydrochloride 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1184980-60-6
:852443-99-3
Formula:C₈H₈ClF₄N
MDL Number:MFCD11226541
:229.61 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together