3-Fluoro-4-(hydroxymethyl)phenylboronic acid pinacol ester
Catalog # APOSPC510176-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:252.09 g/mol
Storage Temperature:Ambient
CAS Number:1082066-29-2
Formula:C₁₃H₁₈BFO₃
MDL Number:MFCD18732617