Order Entry
Portugal
ContactUsLinkComponent
4-(3-Chloro-5-trifluoromethyl-pyridin-2-ylmethyl)-piperazine-1-carboxylic acid tert-butyl ester
4-(3-Chloro-5-trifluoromethyl-pyridin-2-ylmethyl)-piperazine-1-carboxylic acid tert-butyl ester
Catalog # APOSPC446039-100MG
Supplier:  Apollo Scientific
CAS Number:  
4-(3-Chloro-5-trifluoromethyl-pyridin-2-ylmethyl)-piperazine-1-carboxylic acid tert-butyl ester
Catalog # APOSPC446039-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :379.805 g/mol
Storage Temperature:Ambient
CAS Number:1311278-11-1
Formula:C₁₆H₂₁ClF₃N₃O₂
MDL Number:MFCD19981199