Order Entry
Portugal
ContactUsLinkComponent
2-(4-Fluoro-phenylsulphanylmethyl)-pyrrolidine-1-carboxylic acid tert-butyl ester
2-(4-Fluoro-phenylsulphanylmethyl)-pyrrolidine-1-carboxylic acid tert-butyl ester
Catalog # APOSPC408295-500MG
Supplier:  Apollo Scientific
CAS Number:  
2-(4-Fluoro-phenylsulphanylmethyl)-pyrrolidine-1-carboxylic acid tert-butyl ester
Catalog # APOSPC408295-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :311.415 g/mol
Storage Temperature:Ambient
CAS Number:1353945-63-7
Formula:C₁₆H₂₂FNO₂S
MDL Number:MFCD21098435