(2,4-Dimethoxybenzyl)methylamine 97%
Catalog # APOSOR945146-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:181.232 g/mol
Storage Temperature:Ambient
CAS Number:102503-23-1
Formula:C₁₀H₁₅NO₂
MDL Number:MFCD04633426