Order Entry
Portugal
ContactUsLinkComponent
(3-Chloropropyl)sulphamoyl chloride 95%
(3-Chloropropyl)sulphamoyl chloride 95%
Catalog # APOSOR923366-1G
Supplier:  Apollo Scientific
CAS Number:  
(3-Chloropropyl)sulphamoyl chloride 95%
Catalog # APOSOR923366-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

About this item

 :
 :192.064 g/mol
Storage Temperature:Ambient
CAS Number:42065-72-5
Formula:C₃H₇Cl₂NO₂S
MDL Number:MFCD14529128