4-(Piperidin-4-yl)benzoic acid hydrochloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:149353-84-4
:
Formula:C₁₂H₁₅NO₂·ClH
MDL Number:MFCD08460726
:241.714 g/mol
Storage Temperature:Ambient
Frequently Bought Together