N-[(2,4-Dimethoxyphenyl)methyl]-1,2,4-thiadiazol-5-amine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1063733-41-4
:
Formula:C₁₁H₁₃N₃O₂S
MDL Number:MFCD20784227
:251.305 g/mol
Storage Temperature:Ambient
Frequently Bought Together