Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Portugal
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
[6-(Pyrrolidin-1-yl)pyridin-2-yl]methanol
[6-(Pyrrolidin-1-yl)pyridin-2-yl]methanol 97%
[6-(Pyrrolidin-1-yl)pyridin-2-yl]methanol 97%
[6-(Pyrrolidin-1-yl)pyridin-2-yl]methanol 97%
APOSOR12404-250MG
:
Apollo Scientific
:
[6-(Pyrrolidin-1-yl)pyridin-2-yl]methanol 97%
APOSOR12404-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
178.231 g/mol
:
44…46.5 °C
:
Ambient
:
2811
:
6.1,III
:
GHS07
:
868755-48-0
:
C₁₀H₁₄N₂O
:
MFCD09702357