endo-BCN-PEG3-acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1807501-82-1
:
Formula:C₂₀H₃₁NO₇
MDL Number:MFCD31692198
Molecular Weight:397.462 g/mol
:GHS07
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together