
LR white acrylic resins, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:94188-59-7
:
MDL Number:MFCD00131541
:M-Clarity™ quality level = MQ200
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Refractive Index
at 25 ° C: 1.4592 (white light 20 deg C):Conforms - Cure Time
at 25 deg C (Benzoyl Peroxide 1% w.v):≤20 min
Specification Test Results
Frequently Bought Together