Restriction enzymes, EcoRV, HC
Catalog # PROMR435410KU
Supplier: Promega
CAS Number:
Specifications
- Concentration:40 - 80 U/µl
- Pk:10.000 EU
- Source:E. coli J62 pLG74
- Description:EcoRV, HC
- Seq:03
- Enzyme name:EcoRV
- Buffer activity:% Activity (37°C) in 4 CORE® Buffers A: 10…25% – B: 25…50% – C: 50…75% – D: 1; MULTI-CORE™ Buffer 1
- Enzyme assay temperature:37°C
Specifications
About this item
These EcoRV restriction enzyme recognises and cut GAT˅ATC and CTA^TAG at 37 °C in its own unique buffer.
- Capable of digesting DNA in 15 minutes or less
- Active and capable of digestion directly in GoTaq® Green Master Mix
- Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications
- Highy concentrated
Storage buffer: 10 mM Tris-HCl (pH 7,4), 50 mM NaCl, 0,1 mM EDTA, 1 mM DTT, 0,5 mg/ml BSA, 50% glycerol.
Source: Escherichia coli J62 pLG74.
Delivery information: Kit includes 1×1 ml buffer D 10X buffer, 1×150 µl bovine serum albumin (acetylated), 1×2000 units EcoRV and 1×0,25 ml MULTI-CORE™ 10X buffer.