Baths for oil testing, BCA6E and BCA9E
Supplier: SUMINISTROS GRUPO ESPER S L
Certificates
About this item
Digital thermostatic baths for oil testing, controlled by microprocessor. Aluminium block with six or nine holes, specially designed for edible oil testing.
- Digital backlit LCD display showing the selected and actual temperature, and timer (0 to 120 min.)
- Display shows high temperature on the tank, giving protection from burns
- Automatic restart after unexpected shut down keeping the same parameters as before the incident. An incident sign appears on the display
- Memory of the last settings
- Special stainless steel alloy heater is resistant to corrosion and high temperatures
- The unit will cut out if there is insufficient liquid or if the temperature exceeds the maximum temperature
- Push-button Membrane control
Stainless steel inner tank and metallic housing are protected with epoxy paint which has high resistance to aggressive surroundings.
Certifications: IP Protection class: IP 52
Frequently Bought Together