Silver(I) hexafluorophosphate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:26042-63-7
:
Formula:AgF₆P
MDL Number:MFCD00003415
:252.83
ADR:8,II
:M-Clarity™ quality level = MQ100
EINECS:247-428-6
UN:3260
Frequently Bought Together