Bismuth sodium trioxide 85%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12232-99-4
:
Formula:BiNaO₃
MDL Number:MFCD00003474
Merck Index:12,08728
:279.97 g/mol
EINECS:235-455-6
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Yellow to brown Powder
- Titration Iodometric:≥80 %
- Chloride (Cl):≤20 ppm
- Nitrate (NO3):≤40 ppm
- Manganese (Mn):≤5 ppm
Specification Test Results
Frequently Bought Together