Tripropylene glycol diacrylate ≥90.0% (by GC) stabilised
Supplier: TCI
Certificates
About this item
Synonyms: Tri(propyleneglycol)diacrylate
CAS Number:42978-66-5
:
Formula:C₁₅H₂₄O₆
MDL Number:MFCD00064598
:Stabilised with 4-methoxyphenol (MEHQ)
Molecular Weight:300.35 g/mol
Flash Pt:172°C
EINECS:256-032-2
UN:3082
ADR:9 III
:GHS07
:GHS09
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 90.0 % (total of isomer)
Specification Test Results
Frequently Bought Together