
Triphenylsulphonium bromide ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:3353-89-7
:
Formula:C₁₈H₁₅BrS
MDL Number:MFCD00428762
:343.28 g/mol
Melting Pt:293°C
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together