


L-Selenomethionine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:3211-76-5
:
Formula:C₅H₁₁NO₂Se
MDL Number:MFCD00037210
:196.11 g/mol
Merck Index:14,08441
UN:3283
ADR:6.1,III
:GHS06
:GHS08
:GHS09
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to Light yellow powder to crystaline
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Specific rotation [a]20/D:+15.0 to +20.0 deg (C=0.5, 2mol/L HCl)
Specification Test Results
Frequently Bought Together