Sucrose fatty acid ester
: TCI
:
:MFCD00148531
- Appearance:White to Light yellow to Light orange powder to crystal
- Free Sucrose:max. 1.0 %
- Water:max. 4.0 %
- Ignition residue (Sulfate):max. 2.0 %
- Acid Value:max. 6.0
Frequently Bought Together