RuCl[(S,S)-Tsdpen](mesitylene) ≥90% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:174813-81-1
:
Formula:C₃₀H₃₃ClN₂O₂RuS
MDL Number:MFCD12545952
:622.19 g/mol
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (HPLC):min. 90 area%
Specification Test Results
Frequently Bought Together