Order Entry
Portugal
ContactUsLinkComponent
 
Irritant
DL-Phenylephrine hydrochloride ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAP0397-25G
Supplier:  TCI
CAS Number:  
Irritant
DL-Phenylephrine hydrochloride ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAP0397-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    142.0 to 146.0 deg-C
  • Solubility in Water:
    almost transparency

Specification Test Results

About this item

 :
 :203.67 g/mol
EINECS:205-835-6
 :GHS07
CAS Number:154-86-9
Formula:C₉H₁₃NO₂
MDL Number:MFCD00070603