4'-Methyl-4-pentylbiphenyl ≥98.0% (by GC)
: TCI
:64835-63-8
:
:C₁₈H₂₂
:MFCD11053476
:238.37 g/mol
:265-248-6
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:47.0 to 51.0 deg-C
Frequently Bought Together