1-Methyl-1H-pyrazole-3-carboxylic acid ≥97.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:25016-20-0
:
Formula:C₅H₆N₂O₂
MDL Number:MFCD00464254
:126.12 g/mol
:GHS07
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Neutralization titration):min. 97.0 %
- Melting point:153.0 to 157.0 deg-C
Specification Test Results
Frequently Bought Together