Order Entry
Portugal
ContactUsLinkComponent
4-Mercaptohydrocinnamic acid ≥95.0% (by HPLC, titration analysis)
Irritant
4-Mercaptohydrocinnamic acid ≥95.0% (by HPLC, titration analysis)
Catalog # TCIAM0917-1G
Supplier:  TCI
CAS Number:  
Irritant
4-Mercaptohydrocinnamic acid ≥95.0% (by HPLC, titration analysis)
Catalog # TCIAM0917-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (HPLC):
    min. 95.0 area%
  • Purity (Iodometric Titration):
    min. 95.0 %
  • Melting point:
    118.0 to 122.0 deg-C
  • Solubility in hot Methanol:
    very faint turbidity

Specification Test Results

About this item

 :
 :182.24 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:63545-55-1
Formula:C₉H₁₀O₂S
MDL Number:MFCD00060121