Order Entry
Portugal
ContactUsLinkComponent
2-tert-Butyl-6-methylaniline ≥98.0% (by GC, titration analysis)
Irritant
2-tert-Butyl-6-methylaniline ≥98.0% (by GC, titration analysis)
Catalog # TCIAM0763-5ML
Supplier:  TCI
CAS Number:  
Irritant
2-tert-Butyl-6-methylaniline ≥98.0% (by GC, titration analysis)
Catalog # TCIAM0763-5ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 mL
  • Seq:
    2210

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Brown clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :163.26 g/mol
Boiling Pt:250°C
EINECS:236-044-4
 :GHS07
CAS Number:13117-94-7
Formula:C₁₁H₁₇N
MDL Number:MFCD00025208