
4,4'-(Hexafluoroisopropylidene)diphthalic anhydride ≥99.0% (by GC), purified by sublimation
Supplier: TCI
Certificates
About this item
CAS Number:1107-00-2
:
Formula:C₈H₄O₃
MDL Number:MFCD00039143
:444.25 g/mol
EINECS:214-170-0
UN:3261
ADR:8,III
:GHS05
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 99.0 %
- Purity (Morpholine Method):min. 98.0 %
- Melting point:243.0 to 249.0 deg-C
Specification Test Results
Frequently Bought Together