2,3,6,7,10,11-Hexamethoxytriphenylene ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:808-57-1
:
Formula:C₂₄H₂₄O₆
MDL Number:MFCD00075571
:408.45 g/mol
Specification Test Results
- Appearance:Purple to Dark purple to Dark red powder to crystal
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together