L-Glutamic acid diethyl ester hydrochloride ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1118-89-4
:
Formula:C₉H₁₈ClNO₄
MDL Number:MFCD00012509
:239.696 g/mol
EINECS:214-270-4
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
- Melting point:110.0 to 117.0 deg-C
- Specific rotation [a]20/D:+21.0 to +24.0 deg (C=3.6, EtOH)
Specification Test Results
Frequently Bought Together