
3-Formyl-6-methylchromone ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:42059-81-4
:
Formula:C₁₁H₈O₃
MDL Number:MFCD00138943
:188.1794 g/mol
:GHS07
Specification Test Results
- Appearance:White to Yellow to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 97.0 %
- Melting point:170.0 to 174.0 deg-C
Specification Test Results
Frequently Bought Together