
3-(4-Fluorophenyl)propionic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:459-31-4
:
Formula:C₉H₉FO₂
MDL Number:MFCD00060327
:168.17 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:87.0 to 91.0 deg-C
Specification Test Results
Frequently Bought Together