Order Entry
Portugal
ContactUsLinkComponent
 
 
Lithium formate monohydrate ≥98.0% (by titrimetric analysis)
 :  TCI
 
 
Lithium formate monohydrate ≥98.0% (by titrimetric analysis)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Nonaqueous Titration):
    min. 98.0 % (calcd.on anh.substance)
  • Solubility in Water:
    almost transparency
  • Water:
    25.0 to 27.0 %

 

Frequently Bought Together