Dimethyl-7-oxabicyclo[2.2.1]hepta-2,5-diene-2,3-dicarboxylate ≥95.0% (by GC)
: TCI
: 7-Oxabicyclo[2.2.1]hepta-2,5-diene-2,3-dicarboxylic Acid Dimethyl Ester, Dimethyl 3,6-Epoxycyclohexa-1,4-diene-1,2-dicarboxylate, 3,6-Epoxycyclohexa-1,4-diene-1,2-dicarboxylic Acid Dimethyl Ester
:1829-60-3
:
:C₁₀H₁₀O₅
:210.19 g/mol
:120°C
:Refrigerator
- Environmentally Preferable:
- Appearance:Light yellow to Yellow to Orange clear liquid
- Purity (GC):min. 95.0 %
- Specific gravity (20/20):1.2700 to 1.2740
- Refractive index n20/D:1.4940 to 1.4980
Frequently Bought Together