Order Entry
Portugal
ContactUsLinkComponent
(S,S)-(+)-2,3-Dimethoxy-1,4-bis(dimethylamino)butane ≥97.0% (by titrimetric analysis)
Irritant
(S,S)-(+)-2,3-Dimethoxy-1,4-bis(dimethylamino)butane ≥97.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
Irritant
(S,S)-(+)-2,3-Dimethoxy-1,4-bis(dimethylamino)butane ≥97.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light orange to Yellow clear liquid
  • Purity (GC):
    min. 90.0 %
  • Optical purity (GC):
    min. 98.0 ee%
  • Purity (Nonaqueous Titration):
    min. 97.0 %

Specification Test Results

Related Information

Frequently Bought Together