Cyclanilide
1 g
Catalog # TCIAC3595-1G
Supplier: TCI
Jump to:
Product Details & Documents
Synonyms: 1-[(2,4-Dichlorophenyl)carbamoyl]cyclopropanecarboxylic acid
- CAS Number:113136-77-9
- Formula:C₁₁H₉Cl₂NO₃
- MDL Number:MFCD03792791
- Molecular Weight:274.10 g/mol
- Melting Pt:196°C
- EINECS:419-150-7
- UN:2811
- ADR:6.1,III
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:195.0 to 199.0 deg-C
Specifications
- Pk:1 g



