
2-Chloro-p-phenylenediamine ≥98.0% (by GC, titration analysis)
TCIAC3054-25G
: TCI
:
- Pk:25 g
- Seq:3308
- Appearance:White to Gray to Brown powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:63.0 to 67.0 deg-C
:
:142.59 g/mol
:210-441-2
:GHS07
:615-66-7
:C₆H₇ClN₂
:MFCD00128931