Order Entry
Portugal
ContactUsLinkComponent
Charcoal activated catalyst for oxidation using molecular oxygen
Charcoal activated catalyst for oxidation using molecular oxygen
Catalog # TCIAC2194-100G
Supplier:  TCI
CAS Number:  
Charcoal activated catalyst for oxidation using molecular oxygen
Catalog # TCIAC2194-100G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Appearance:
    Black powder to crystal
  • Drying loss:
    max. 10.0 %
  • Median size:
    30.0 to 60.0 micro_m

Specification Test Results

About this item

 :
 :12.01 g/mol
Merck Index:14,01807
EINECS:231-153-3
CAS Number:7440-44-0
 :64365-11-3
Formula:C
MDL Number:MFCD00133992