Carbon nanotube multi-walled, Length: 5-15 µm, Ø 40-60 nm
Supplier: TCI
Certificates
About this item
Synonyms: CNT
CAS Number:308068-56-6
:
Formula:C
MDL Number:MFCD00133992
:12.011 g/mol
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Black powder to crystal
- Purity (TPO Method):min. 97 %
- Diameter:40 to 60 nm
- Length:5 to 15 micro_m
Specification Test Results
Frequently Bought Together