
Ethyl-2,4-dihydroxy-6-methylbenzoate ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:2524-37-0
:
Formula:C₁₀H₁₂O₄
MDL Number:MFCD00010821
:196.20 g/mol
:GHS07
Specification Test Results
- Appearance:White to Gray to Brown powder to crystal
- Purity (GC):min. 95.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:131.0 to 134.0 deg-C
Specification Test Results
Frequently Bought Together